Learn More
FAT2 Antibody (8C5), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-54371APC
Product Code. 18696937
  • € 617.00 / 100µL
Description
FAT2 Monoclonal antibody specifically detects FAT2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FAT2
Monoclonal
APC
protocadherin Fat 2, CDHF8, CDHR9, FAT tumor suppressor homolog 2 (Drosophila), HFAT2, MEGF1
Purified recombinant human FAT2 protein. (Uniprot: Q9NYQ8)
100 μg
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
8C5
PBS
Mouse
Protein A or G purified
RUO
2196
Store at 4°C in the dark.