Learn More
Invitrogen™ HSF1 Monoclonal Antibody (10H8), PE
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA545130
Product Code. 17848792
  • € 433.00 / 100µg
Description
1 μg/mL of MA5-45130 was sufficient for detection of HSF1 in 20 μg of heat shocked HeLa cell lysate by ECL immunoblot analysis using Goat anti-rat IgG: HRP as the secondary antibody Detects approximately 85kDa (unstressed cell lysates), andapproximately 95kDa (heat shocked cell lysates).
All organisms respond to elevated temperatures and a variety of environmental stresses by rapid synthesis of heat shock RNAs and proteins. The regulation of heat shock gene transcription is mediated by the transcriptional activator, heat shock factor (HSF), which binds to heat shock response elements (HSEs). These HSEs are found as three repeats of a 5-nucleotide {nGAAn} module, arranged in alternating orientation and present upstream of all heat shock genes. The HSEs are highly conserved among species yet HSF purified from yeast, Drosophila and human have different molecular weights and the proteins do not show significant immunological cross reaction. Two HSFs have been identified in human cells, HSF1 and HSF2, which bind to the same HSEs and have 38% sequence identity. These factors are activated by distinct stimuli, HSF1 is responsive to classical stress signals such as heat, heavy metals and oxidative reagents, whereas HSF2 is activated during hemin-mediated differentiation of human erythroleukemia cells. HSF1 exists constitutively in the cytoplasm and the nucleus of unstressed cells as a monomer which lacks DNA binding activity. Through an unknown signal generated during stress, HSF1 becomes activated to a nuclear localized, trimeric state which binds to DNA. The phosphorylation of HSF1 is necessary for maximal transcription of heat shock genes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HSF1
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.2
P38532, Q00613, Q08DJ8
Hsf1
Purified recombinant mouse HSF1 protein, with epitope mapping to amino acids 378-395.
100 μg
Primary
Human, Mouse, Rat, Rabbit, Monkey, Bovine, Hamster, Guinea Pig
Antibody
IgG1
ELISA, Gel Shift, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry
10H8
PE
Hsf1
AA960185; heat shock factor 1; heat shock factor 2; Heat shock factor protein 1; Heat shock transcription factor 1; heat shock transcription factor 1 alpha isoform; heat shock transcription factor 1 beta isoform; heat shock transcription factor 1 gammaalpha isoform; heat shock transcription factor 1 gammabeta isoform; HSF; HSF 1; Hsf1; Hsf1alpha; Hsf1beta; Hsf1gammaalpha; Hsf1gammabeta; HSTF 1; HSTF1
Rat
Protein G
RUO
100350887, 100762942, 101787582, 15499, 3297, 506235, 79245
4°C