missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1, p Ser326 Antibody (SR2246), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21520-100ul
This item is not returnable.
View return policy
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Specifications
| HSF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| SR2246 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| A synthesized peptide derived from human phospho-HSF1 [Ser326] (Uniprot #: Q00613 ) | |
| 100 μg | |
| Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
| 3297 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction