missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF2 Antibody (3E2), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-96434G
This item is not returnable.
View return policy
Description
HSF2 Monoclonal antibody specifically detects HSF2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunofluorescence
Specifications
| HSF2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig | |
| Purified |
| Western Blot, Immunofluorescence | |
| 300 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
| Purified recombinant mouse HSF2 protein | |
| 0.1 mL | |
| Cellular Signaling, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 3298 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction