missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF2BP Monoclonal Antibody (OTI2A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810898S
This item is not returnable.
View return policy
Description
HSF2BP Monoclonal antibody specifically detects HSF2BP in Human samples. It is validated for Western BlotSpecifications
| HSF2BP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50991 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A1 | |
| Unconjugated | |
| Cct4 | |
| 4932437G14Rik; heat shock factor 2-binding protein; heat shock transcription factor 2 binding protein; Hsf2bp; RGD1565879 | |
| Full length human recombinant protein of human CCT4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction