missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF4 Antibody (2E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 205.00 - € 552.00
Specifications
| Antigen | HSF4 |
|---|---|
| Clone | 20000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670656
|
Novus Biologicals
NBP2-37499-0.025ml |
0.025 mL |
€ 205.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18165049
|
Novus Biologicals
NBP2-37499 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSF4 Monoclonal specifically detects HSF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| HSF4 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| HSF4 | |
| IgG1 | |
| Affinity Purified |
| 20000000 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| 3299 | |
| Purified recombinant fragment of human HSF4 expressed in E. Coli. | |
| Primary | |
| 53 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title