missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA116966
This item is not returnable.
View return policy
Description
PA1-16966 is specific for the ~84 kDa Hormone-Sensitive Lipase protein. A non-specific band may also be detected at ~89 kDa. Bands at ~102, 113, and 127 kDa may also be seen in the testis of sexually immature rats.Suggested positive control: human adipose protein.
Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
Specifications
| HSL | |
| Polyclonal | |
| Unconjugated | |
| LIPE | |
| 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| 16890, 25330, 3991 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P15304, P54310, Q05469 | |
| LIPE | |
| A fusion protein containing an internal portion of the rat HSL protein (between residues 699-908). | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction