missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP20 Monoclonal Antibody (C9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543765
This item is not returnable.
View return policy
Description
HSP20 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Heat Shock Protein 20 (HSP20), encoded by the HSPB6 gene, is a member of the small heat shock protein (sHSP) family, prominently involved in cellular stress response and maintenance of cytoskeletal integrity. HSP20 is ubiquitously expressed across various tissues, including cardiac and smooth muscle, where its roles are vital in modulating stress resilience and muscle function. It exerts protective effects against apoptosis and oxidative stress by interacting with cell signaling pathways that enhance cellular survival and protein stabilization. In cardiac muscle, HSP20 is known for its cardioprotective actions during ischemic injury, helping preserve myocardial function by inhibiting hypertrophic signaling pathways and promoting muscle relaxation. Studies highlight its involvement in vasodilation processes, as HSP20 interacts with myosin to regulate smooth muscle contraction and dilatation. Beyond its role in muscle physiology, HSP20 has been investigated for its potential therapeutic applications in cardiovascular diseases and metabolic disorders, where its expression and function could be pivotal in disease modulation and treatment strategies.
Specifications
| HSP20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P97541 | |
| HSPB6 | |
| Recombinant protein Heat Shock Protein Beta 6. The antigen corresponds to amino acid range 1-162 of the target protein. | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C9 | |
| Unconjugated | |
| HSPB6 | |
| AA387366; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; HSPB6; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 192245 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction