missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody (8A7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 204.00
Specifications
| Antigen | HSP27 |
|---|---|
| Clone | 8A7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Knockout Validated |
| Applications | Western Blot |
Description
HSP27 Monoclonal specifically detects HSP27 in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Hamster, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| HSP27 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| P04792 | |
| 3315 | |
| HSP27 peptide | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 8A7 | |
| Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| HSPB1 | |
| IgG1 κ | |
| Protein G purified | |
| Detects approx 25kDa or approx 27kDa. Recognizes HSP25 and HSP27, cross reacts with alpha B crystallin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title