missing translation for 'onlineSavingsMsg'
Learn More

HSP27 Mouse, Clone: G3.1, Novus Biologicals™

Product Code. 18165424 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18165424 0.02 mg 0.02mg
18705723 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18165424 Supplier Novus Biologicals Supplier No. NBP2329720.02MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSP27 Monoclonal specifically detects HSP27 in Human, Mouse, Rat, Chicken, Chimpanzee, Monkey, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP27
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G3.1
Conjugate Unconjugated
Dilution Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P04792
Gene Alias 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Gene Symbols HSPB1
Host Species Mouse
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Molecular Weight of Antigen 27 kDa
Purification Method Protein A purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3315
Test Specificity This MAb reacts specifically with heat shock protein HSP27 in human and monkey tissues and cell lines such as MCF-7. HSP27, also referred to as the Estrogen-Regulated 24K protein and HSP28, is one of several small heat shock proteins produced by all organisms studied. HSP27 synthesis is induced by elevated temperature, as well as by estrogen in hormone responsive cells. Interestingly, human HSP27 also shares greater than 50% homology with low molecular weight Drosophila HSPs and mammalian alpha-crystalline lens protein. Because of the estrogen responsive nature of HSP27, this protein has been studied extensively in human estrogen responsive tissues such as cervix, endometrium and breast tissue. Therefore HSP27 may be useful in classifying various hormone sensitive tumors.
Target Species Human, Mouse, Rat, Chicken, Primate, Sheep
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.