missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Mouse, Clone: SPM252, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32865-0.1MG
This item is not returnable.
View return policy
Description
HSP27 Monoclonal specifically detects HSP27 in Human, Mouse, Rat, Chicken, Chimpanzee, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HSP27 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Mouse | |
| 27 kDa | |
| 0.1 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| 3315 | |
| Human, Mouse, Rat, Chicken, Primate, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM252 | |
| Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| P04792 | |
| HSPB1 | |
| Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells. | |
| Protein A purified | |
| RUO | |
| Primary | |
| It recognizes a 24-27kDa estrogen-regulated protein, identified as heat shock protein 27 (hsp27). Hsp27 was recently found to be identical to the estrogen-induced p29 and 24K protein. About 50% of breast carcinomas are positive for hsp27 especially those that are also positive for estrogen and/or progesterone receptor. HSP27 has also been implicated in drug resistance in cancer cells. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction