missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27, p Ser78 Antibody (2079A), Alexa Fluor™ 750, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB9355AF750
This item is not returnable.
View return policy
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry.
Specifications
| HSP27 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HSPB1 | |
| Phosphopeptide containing the human HSP27 S78 site, Accession No. P04792 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 2079A | |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Rabbit | |
| Protein A or G purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| 3315.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction