missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP27 Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143536
This item is not returnable.
View return policy
Description
HSP27 Polyclonal Antibody for Western Blot, ICC/IF
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. HSPB1 (heat shock protein beta-1 or HSP27) is a small heat shock protein which functions as a molecular chaperone that maintains denatured proteins in a folding-competent state. It plays a role in stress resistance and actin organization. Through its molecular chaperone activity, HSP27 regulates numerous biological processes including the phosphorylation and the axonal transport of neurofilament proteins. Mutations in the gene can result in Charcot-Marie-Tooth disease 2F and Neuronopathy distal hereditary motor 2B.
Specifications
| HSP27 | |
| Polyclonal | |
| Unconjugated | |
| HSPB1 | |
| 27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25 | |
| Chicken | |
| 500 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgY |
| Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| PBS with 5mM sodium azide | |
| P04792, P42930 | |
| HSPB1 | |
| Recombinant full length human HSP27 expressed in and purified from E. coli. | |
| RUO | |
| 24471, 3315 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction