Learn More
Invitrogen™ Hsp31p Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142906
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Catalyzes the conversion of methylglyoxal (MG) to D-lactate in a single glutathione (GSH)-independent step. May play a role in detoxifying endogenously produced glyoxals (PubMed:24302734). Involved in protection against reactive oxygen species (ROS) (PubMed:17395014). Important for viability in stationary phase. May negatively regulate TORC1 in response to nutrient limitation (PubMed:24706893).
Specifications
| Hsp31p | |
| Polyclonal | |
| Unconjugated | |
| HSP31 | |
| HSP31 | |
| Peptide with sequence C-ETGKFGWDEHSLAKD (aa 50-64). | |
| 100 μg | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q04432 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 852146 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |