missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40/DNAJB1 Antibody (3B9.E6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12876AF405
This item is not returnable.
View return policy
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HSP40/DNAJB1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DNAJB1 | |
| Recombinant Protein HSP40 (Hdj1) | |
| 100 μL | |
| Primary | |
| Detects approx 40kDa. Does not cross-react with HDJ2 or YDJ1. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3B9.E6 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
| Mouse | |
| Protein G purified | |
| DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| 3337 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction