missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP40 Monoclonal Antibody (2A7.H6), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545221
This item is not returnable.
View return policy
Description
0.5 μg/mL of MA5-45221 was sufficient for detection of 50 ng YDJ1 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Yeast specific. Does not cross react with Human, Mouse or Rat.
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
Specifications
| HSP40 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative | |
| Mouse | |
| Protein G | |
| RUO | |
| Yeast | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 2A7.H6 | |
| FITC | |
| 0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 | |
| Full length protein HSP40 (YDJ1). | |
| 100 μg | |
| Primary | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction