missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp40/YDJ1 Antibody (1G10.H8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12879FR
This item is not returnable.
View return policy
Description
Hsp40/YDJ1 Monoclonal specifically detects Hsp40/YDJ1 in Yeast samples. It is validated for Western Blot, ELISA.
Specifications
| Hsp40/YDJ1 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| YDJ1 | |
| Full length protein yeast HSP40 (YDJ1) | |
| 100 μL | |
| 855661 | |
| Yeast | |
| Purified |
| Western Blot, ELISA | |
| 1G10.H8 | |
| Western Blot, ELISA | |
| MAS5, YDJ1 Ydj1p | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 40kDa. Yeast specific product. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction