missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp47 Antibody (1C4-1A6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22400B
This item is not returnable.
View return policy
Description
Hsp47 Monoclonal antibody specifically detects Hsp47 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Hsp47 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1C4-1A6 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin | |
| Human HSP47, full length | |
| 0.2 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 871 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction