missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody (LK2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34671CL1
This item is not returnable.
View return policy
Description
HSP60 Monoclonal antibody specifically detects HSP60 in Bacteria,Chicken,E. coli,Guinea Pig,Hamster,Human,Invertebrate,Monkey,Mouse,Parasite,Plant,Rat,S. pyogenes,Yeast samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| HSP60 | |
| Monoclonal | |
| CoraFluor 1 | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Recombinant human HSP60 protein (Uniprot: P10809 ) | |
| 0.1 mL | |
| Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| 3329 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Simple Western, CyTOF | |
| LK2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bacteria, Chicken, E. coli, Guinea Pig, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction