missing translation for 'onlineSavingsMsg'
Learn More

HSP60 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-12734-0.025mg

Product Code. 18723004

  • € 213.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSP60 Polyclonal antibody specifically detects HSP60 in Human, Bacteria, P. falciparum samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSP60
Polyclonal
Unconjugated
PBS (pH 7.4), 50% Glycerol
Rabbit
Protein A purified
RUO
Primary
Human, Bacteria, P. falciparum
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
1.83 mg/mL
Western Blot 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Recombinant full length PfHSP60
0.025 mg
Apoptosis
3329
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.