missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody, Novus Biologicals™
Description
HSP60 Polyclonal antibody specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | HSP60 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10000-1:20000, Immunohistochemistry 1: 2000-1:5000, Immunocytochemistry/ Immunofluorescence 1:2000-1:5000 |
| Formulation | Supplied as an aliquot of IgY preparation |
| Gene Alias | 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) |
| Host Species | Chicken |
| Immunogen | Recombinant full length human HSP60 expressed in and purified from E. coli |
| Purification Method | IgY purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?