missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp60 (HSPD1) Monoclonal Antibody (OTI3A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800758S
This item is not returnable.
View return policy
Description
Hsp60 (HSPD1) Monoclonal antibody specifically detects Hsp60 (HSPD1) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| Hsp60 (HSPD1) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10809 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3329 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A2 | |
| Unconjugated | |
| HSPD1 | |
| 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409) | |
| Full length human recombinant protein of human HSPD1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction