missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70/HSPA1A Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77455MFV610
This item is not returnable.
View return policy
Description
HSP70/HSPA1A Polyclonal antibody specifically detects HSP70/HSPA1A in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| HSP70/HSPA1A | |
| Polyclonal | |
| Flow Cytometry | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
| A synthetic peptide made to an internal portion of the human Hsp70 protein (between residues 550-600) [Uniprot: P08107] | |
| 0.1 mL | |
| Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 3303 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction