missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 alpha Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77682H
This item is not returnable.
View return policy
Description
HSP90 alpha Polyclonal antibody specifically detects HSP90 alpha in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HSP90 alpha | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| A synthetic peptide made to an N-terminal portion of the human Hsp90A protein (between residues 1-70) [UniProt P07900] | |
| 0.1 mL | |
| Cancer, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones | |
| 3320 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction