missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 beta Antibody (HSP90AB1/3954), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08937AF350
This item is not returnable.
View return policy
Description
HSP90 beta Monoclonal antibody specifically detects HSP90 beta in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| HSP90 beta | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984 | |
| Recombinant human HSP90 beta protein fragment (around aa581-704) (exact sequence is proprietary) (Uniprot: P08238) | |
| 100 μg | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| 3326 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| HSP90AB1/3954 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction