missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 beta Antibody (HSP90AB1/4970), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20590-100ug
This item is not returnable.
View return policy
Description
HSP90 beta Monoclonal antibody specifically detects HSP90 beta in Human, Mouse, Rat, Canine, Chicken, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HSP90 beta | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984 | |
| Recombinant full-length human HSP90 beta protein | |
| 100 μg | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 3326 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HSP90AB1/4970 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction