missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3F11C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60318-1-IG-20UL
This item is not returnable.
View return policy
Description
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).Specifications
| HSP90 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O02705, P07900, P07901, P82995 | |
| HSP90AA1 | |
| HSP90 Fusion Protein Ag3826 | |
| 20 μL | |
| Primary | |
| Pig, Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 3F11C1 | |
| Unconjugated | |
| HSP90AA1 | |
| Heat shock 86 kDa, HSP 86, HSP86, Hsp89, HSP89A, HSP90, HSP90A, HSP90AA1, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, LAP2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 15519, 299331, 3320, 397028 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction