missing translation for 'onlineSavingsMsg'
Learn More

Hsp90A/B Antibody (Hyb-K41220A), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18701442 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18701442 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18701442 Supplier Novus Biologicals Supplier No. NB11096870JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hsp90A/B Monoclonal antibody specifically detects Hsp90A/B in Human, Mouse, Rat, Yeast samples. It is validated for ELISA, Immunohistochemistry, Western Blot (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp90A/B
Applications ELISA, Immunohistochemistry, Western Blot (Negative)
Classification Monoclonal
Clone Hyb-K41220A
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Host Species Mouse
Immunogen Recombinant human HSP90alpha; Specificity mapped to amino acids 291-304
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3320
Target Species Human, Mouse, Rat, Yeast
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.