missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA1B Antibody (HSPA1B/7625), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24190AF532
This item is not returnable.
View return policy
Description
HspA1B Monoclonal antibody specifically detects HspA1B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HspA1B | |
| Monoclonal | |
| Alexa Fluor 532 | |
| FLJ54328, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1B, heat shock 70kDa protein 1B, HSP70.1/HSP70.2, HSP70-1/HSP70-2, HSP70-1B, HSP70-2, HSPA1, HSPA1A | |
| Recombinant fragment (around aa400-600) of human HspA1B protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 3304 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| HSPA1B/7625 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction