missing translation for 'onlineSavingsMsg'
Learn More

HspA1B Antibody (HSPA1B/7627), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24280AF594

Product Code. 18561209

  • € 679.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HspA1B Monoclonal antibody specifically detects HspA1B in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HspA1B
Monoclonal
Alexa Fluor 594
FLJ54328, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1B, heat shock 70kDa protein 1B, HSP70.1/HSP70.2, HSP70-1/HSP70-2, HSP70-1B, HSP70-2, HSPA1, HSPA1A
Recombinant fragment (around aa400-600) of human HspA1B protein (exact sequence is proprietary)
0.1 mL
Cancer
3304
Store at 4°C in the dark.
IgG2b κ
Immunohistochemistry (Paraffin)
HSPA1B/7627
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.