missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPA1B Monoclonal Antibody (C8), Invitrogen™

Product Code. 17930233
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930233 20 μL 20µL
17960463 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17930233 Supplier Invitrogen™ Supplier No. MA551014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This intronless gene encodes a 70 kDa heat shock protein which is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA1B
Applications Western Blot
Classification Monoclonal
Clone C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Hspa1b
Gene Accession No. P0DMV9, Q27965
Gene Alias 68 kDa heat shock protein; Hcp70.1; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kDa protein 1B; Heat shock protein; heat shock protein 1B; heat shock protein 70.1; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; HSP; hsp68; Hsp70; HSP70.1; HSP70.1/HSP70.2; HSP70.2; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-2; Hsp70a1; HSP70I; Hsp72; HSPA1; Hspa1a; HSPA1B; Hspa2; HSX70
Gene Symbols Hspa1b
Host Species Mouse
Immunogen Recombinant Bovine HSPA1B protein, Met1-Asp641 (Accession #Q27965).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282254, 3304
Target Species Bovine, Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.