missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA1L Monoclonal Antibody (OTI3B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812122S
This item is not returnable.
View return policy
Description
HSPA1L Monoclonal antibody specifically detects HSPA1L in Human samples. It is validated for Western BlotSpecifications
| HSPA1L | |
| Monoclonal | |
| 0.5 to 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14061 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10063 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3B5 | |
| Unconjugated | |
| COX17 | |
| heat shock 10kDa protein 1-like; heat shock 70 kDa protein 1-Hom; heat shock 70 kDa protein 1L; Heat shock 70 kDa protein 1-like; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein-like 1; heat shock 70kDa protein 1-like; Heat shock protein; heat shock protein 1-like; Heat shock protein 70-1l; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock-related protein hsc70t; Hsc70t; HSP70.3; HSP70-1L; Hsp70-3; HSP70-HOM; HSP70T; Hspa1l; hum70t; inducible heat shock protein 70; Msh5; Spermatid-specific heat shock protein 70 | |
| Human recombinant protein fragment corresponding to amino acids 2-63 of human COX17 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction