Learn More
HSPA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12797-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody well recognized the endogenous HSPA4 protein in testis and multiple cell lines.
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.Specifications
| HSPA2 | |
| Polyclonal | |
| Unconjugated | |
| HSPA2 | |
| Heat shock 70 kDa protein 2, heat shock 70kDa protein 2, HSP70 2, HSP70 3, HSPA2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15512, 3306, 60460 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14659, P17156, P54652 | |
| HSPA2 | |
| HSPA2 Fusion Protein Ag3539 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |