Learn More
HSPA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12797-1-AP-20UL
 View more versions of this product
Product Code. 16850704
  • € 145.00 / 20µL
Description
This antibody well recognized the endogenous HSPA4 protein in testis and multiple cell lines.
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Specifications
Specifications
HSPA2
Polyclonal
Unconjugated
HSPA2
Heat shock 70 kDa protein 2, heat shock 70kDa protein 2, HSP70 2, HSP70 3, HSPA2
Rabbit
Antigen Affinity Chromatography
RUO
15512, 3306, 60460
-20°C
Liquid
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14659, P17156, P54652
HSPA2
HSPA2 Fusion Protein Ag3539
20 μL
Primary
Human, Mouse, Rat