missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP1-81696-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HspA4 Polyclonal specifically detects HspA4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| HspA4 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| APG2, APG-2, heat shock 70 kDa protein 4, heat shock 70kD protein 4, heat shock 70kDa protein 4, Heat shock 70-related protein APG-2, heat shock protein, 110 kDa, HS24/P52, hsp70, hsp70RY, MGC131852, RY | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3308 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HSPA4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FEELGKQIQQYMKIISSFKNKEDQYDHLDAADMTKVEKSTNEAMEWMNNKLNLQNKQSLTMDPVVKSKEIEAKIKELTSTCSPIISKPKPKVEPPKEEQKNAEQNGPVDG | |
| 25 μL | |
| Primary | |
| Specificity of human HspA4 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering