Learn More
Invitrogen™ HSPA4 Monoclonal Antibody (3A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA519153
Description
Peptide Sequence: MSVVGIDLGF QSCYVAVARA GGIETIANEY SDRCTPACIS FGPKNRSIGA AAKSQVISNA KNTVQGFKRF HGRAFSDPFV EAEKSNLAYD IVQLPTGLTG IKVTYMEEEQ NGPVDGQGDN PGPQAAEQGT DTAVPSDSDK KLPEMDID.
The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activity.
Specifications
| HSPA4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P34932 | |
| HSPA4 | |
| HSPA4 (AAH02526, 1 a.a. ~ 148 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3A11 | |
| Unconjugated | |
| HSPA4 | |
| 70kDa; AI317151; APG2; APG-2; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 4; heat shock 70kD protein 4; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; Heat shock protein; heat shock protein 4; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; HEL-S-5a; HS24/P52; Hsp110; hsp70; hsp70 RY; Hsp70RY; Hspa4; HSPH2; Irp94; ischemia responsive 94 kDa protein; mKIAA4025; RY | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.