missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA6 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97536AF488
This item is not returnable.
View return policy
Description
HspA6 Polyclonal specifically detects HspA6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| HspA6 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 3310 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 488 | |
| heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B' | |
| Produced in rabbits immunized with E. coli-derived Human HspA6 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction