missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA6 Antibody (OTI1H3), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70959PEATT594
This item is not returnable.
View return policy
Description
HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Flow Cytometry.
Specifications
| HspA6 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| HSPA6 | |
| Full length recombinant protein of human HspA6 (NP_002146) produced in HEK293T cells. | |
| 0.1 mL | |
| 3310.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1H3 | |
| Flow Cytometry | |
| heat shock 70 kDa protein 6, Heat shock 70 kDa protein B′, heat shock 70kD protein 6 (HSP70B′), heat shock 70kDa protein 6 (HSP70B′), HSP70B′ | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction