missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (1B5), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-97868-0.025MG
This item is not returnable.
View return policy
Description
HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications
| HSPA8/HSC71/Hsc70 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Electron Microscopy 1:10-1:500, Enzyme Immunoassay | |
| Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 3312 | |
| Pig, Bovine, Hamster, Sheep, Canine, Chicken, Rat, Rabbit, Human, Guinea Pig, Monkey, Mouse | |
| Purified |
| Immunocytochemistry, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry | |
| 1B5 | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| HSPA8 | |
| Native hamster Hsc70 (Hsp73). | |
| 0.025 mg | |
| Primary | |
| Recognizes dog, chicken, guinea pig, pig, rabbit, sheep, and turtle HSC70 (HSP73). | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction