missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPA9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17741404
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17741404 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17741404 Supplier Invitrogen™ Supplier No. PA5120002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: PC-12 cell lysate, MCF-7 cell lysate, Hela cell lysate, NIH/3T3 cell lysate, mouse kidney tissue lysate, mouse brain tissue lysate, HepG2, MCF-7, N2A, SH-SY5Y, mouse testis tissue, mouse brain tissue, mouse prostate tissue. Subcellular Location: Mitochondrion, nucleolus.

The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA9
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene HSPA9
Gene Accession No. P38646, P38647, P48721
Gene Alias 70 kDa heat shock protein; 74kDa; 75 kDa glucose-regulated protein; C3H-specific antigen; catecholamine-regulated protein 40; CRP40; CSA; epididymis secretory sperm binding protein Li 124m; EVPLS; GRP 75; GRP75; GRP-75; heat shock 70 kDa protein 9; heat shock 70 kDa protein 9B; heat shock 70kD protein 9B; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; heat shock 70kDa protein 9B (mortalin-2); Heat shock protein; heat shock protein 9; heat shock protein 9A; heat shock protein cognate 74; heat shock protein family A (Hsp70) member 9; heat shock protein family A (Hsp70) member 9 S homeolog; heat shock protein, 74 kDa, A; heat shock protein, A; HEL-S-124m; Hsc74; HSP; Hsp74; Hsp74a; hspa9; hspa9.S; Hspa9a; hspa9-a; hspa9b; hspa9-b; hypothetical protein; I79_009139; MGC4500; mortalin; mortalin, perinuclear; mortalin2; mortalin-2; mot; MOT2; Mot-2; Mthsp70; mt-HSP70; MTHSP75; p66 MOT; p66-mortalin; pbp74; Peptide-binding protein 74; RCJMB04_2m8; SAAN; SIDBA4; stress-70 protein mitochondrial-like protein; stress-70 protein, mitochondrial; Stress-70 protein, mitochondrial-like protein; XELAEV_18020021mg
Gene Symbols HSPA9
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal human Grp75.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15526, 291671, 3313
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.