missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspB7 Antibody (OTI1D11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71833H
This item is not returnable.
View return policy
Description
HspB7 Monoclonal specifically detects HspB7 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HspB7 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| HSPB7 | |
| Full length recombinant protein of human HspB7 (NP_055239) produced in HEK293T cells. | |
| 0.1 mL | |
| 27129.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1D11 | |
| Western Blot | |
| Cardiovascular heat shock protein, CVHSP, DKFZp779D0968, FLJ32733, heat shock 27kDa protein family, member 7 (cardiovascular), heat shock protein beta-7, HspB7, member 7 (cardiovascular) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction