missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPB8/HSP22 Antibody (OTI1E3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71828CL1
This item is not returnable.
View return policy
Description
HSPB8/HSP22 Monoclonal antibody specifically detects HSPB8/HSP22 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| HSPB8/HSP22 | |
| Monoclonal | |
| CoraFluor 1 | |
| Alpha-crystallin C chain, CRYAC, E2IG1E2-induced gene 1 protein, H11DHMN2, heat shock 22kDa protein 8, heat shock 27kDa protein 8, heat shock protein beta-8, HMN2, HMN2A, HSP22CMT2L, HspB8, Protein kinase H11, Small stress protein-like protein HSP22 | |
| Full length human recombinant protein of human HSPB8(NP_055180) produced in HEK293T cell. | |
| 0.1 mL | |
| Protein Kinase | |
| 26353 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI1E3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction