missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPB9 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5144846

Product Code. 18016591

  • € 604.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TGQQQLDVRD PERVSYRMSQ KVHRKMLPSN LSPTAMTCCL TPSGQLWVRG QCVALALPEA QTGPSPRLGS LGSKASNLTR Antigen sequence identity: Mouse 55%; Rat 48%.

HSPB9 gene ontology (GO) annotation includes protein binding and nucleus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSPB9
Polyclonal
Unconjugated
HSPB9
1700007H20Rik; Cancer/testis antigen 51; CT51; Heat shock protein; Heat shock protein beta-9; heat shock protein family B (small) member 9; heat shock protein, alpha-crystallin-related, B9; HSP; HSPB9; small heat shock protein B9
Rabbit
Affinity chromatography
RUO
94086
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
IgG
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BQS6
HSPB9
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 80-159. Recombinant protein control fragment (Product #RP-109783).
100 μL
Primary
Human
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.