missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPB9 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 185.00 - € 405.00
Specifications
| Antigen | HSPB9 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17980533
|
Invitrogen™
PA5145460 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17968592
![]() |
Invitrogen™
PA5145459 |
200 μL |
€ 405.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSPB9 gene ontology (GO) annotation includes protein binding and nucleus.Specifications
| HSPB9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9BQS6, Q9DAM3 | |
| 75482, 94086 | |
| Recombinant Human HSPb9 protein, Met1-Thr158 (Accession #Q9BQS6). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 1700007H20Rik; Cancer/testis antigen 51; CT51; Heat shock protein; Heat shock protein beta-9; heat shock protein family B (small) member 9; heat shock protein, alpha-crystallin-related, B9; HSP; HSPB9; small heat shock protein B9 | |
| HSPB9 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| HSPB9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title