missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspBP1 Antibody (OTI1D5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71761PCP
This item is not returnable.
View return policy
Description
HspBP1 Monoclonal specifically detects HspBP1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications
| HspBP1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| HSPBP1 | |
| Full length recombinant protein of human HspBP1 (NP_036399) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI1D5 | |
| Flow Cytometry | |
| FES1, Heat shock protein-binding protein 1, Hsp70 binding protein 1, hsp70 interacting protein, hsp70-binding protein 1, Hsp70-binding protein 2, Hsp70-interacting protein 1, Hsp70-interacting protein 2, HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1, HSPBP, HspBP1, hspBP2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 23640.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction