Learn More
PDGFR alpha Antibody (16A1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-44581PE
Product Code. 18223923
  • ONLINE EXCLUSIVE PRICE
    € 540.00
    € 510.30 / 0.10mL
    Save € 29.70
    5% Off
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDGFR alpha
Monoclonal
PE
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Mouse
Protein A purified
RUO
Primary
The mouse monoclonal antibody 16A1 recognizes CD140a PDGF-RA, the 170 kDa alpha chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative or anti-proliferative roles in various tumours. HLDA VI.; WS Code E022
Store at 4C in the dark.
IgG1
Flow Cytometry
16A1
Flow Cytometry
PDGFRA
CD140a-transfected NIH 3T3 cells
0.1 mL
Phospho Specific, Protein Kinase
5156