Learn More
HSPD1 Mouse anti-Human, Clone: OTI1B3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800757
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Two pseudogenes, both located on chromosome 8, have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.Specifications
| HSPD1 | |
| Monoclonal | |
| Unconjugated | |
| HSPD1 | |
| CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3329 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P10809 | |
| HSPD1 | |
| Full length human recombit protein of human HSPD1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.