missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPH1/HSP105 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97291MFV500
This item is not returnable.
View return policy
Description
HSPH1/HSP105 Polyclonal antibody specifically detects HSPH1/HSP105 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HSPH1/HSP105 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| Antigen NY-CO-25, heat shock 105kD beta, heat shock 105kDa protein 1, heat shock 105kDa/110kDa protein 1, Heat shock 110 kDa protein, heat shock protein 105 kDa, HSP105A, HSP105B, HSP105heat shock 105kD alpha, HSP110, KIAA0201DKFZp686M05240, NY-CO-25 | |
| Produced in rabbits immunized with E. coli-derived Human HSPH1/HSP105 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 10808 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction