Learn More
BOLA HT Replacement Inner Parts
Product Code. 10003281
Change view
Click to view available options
Thread Size:
32
45
Diameter (Metric):
3 mm
3.2 mm
5 mm
6 mm
8 mm
9.5 mm
9.52 mm
12.7 mm
14 mm
16 mm
18 mm
18.01 mm
19.05 mm
20 mm
21 mm
22 mm
30 mm
32 mm
Unit Size:
Each
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Diameter (Metric) Unit Size
10003281 45 21 mm Each
15583563 32 5 mm Each
10526132 32 9.52 mm Each
10554592 32 3 mm Each
10718442 32 9.5 mm Each
10718632 32 12.7 mm Each
10514402 32 14 mm Each
10013221 45 18.01 mm Each
15335077 45 19.05 mm Each
10766172 45 20 mm Each
10287211 45 3.2 mm Each
10689021 45 6 mm Each
10043241 45 8 mm Each
10093261 45 14 mm Each
10309141 45 16 mm Each
10477137 45 18 mm Each
10174631 45 22 mm Each
10033511 45 30 mm Each
10227551 45 32 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 10003281 Supplier BOLA Supplier No. D64260
  • ONLINE EXCLUSIVE PRICE
    € 101.85
    € 83.19 / Pack of 1
    Save € 18.66
    18% Off
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
HT Replacement Inner Parts inner parts made of PPS and PTFE max. temperature 250°C
Consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm (not exposed to the medium). Good chemical resistance, the medium is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Diameter (Metric) 21 mm
Autoclavable Autoclavable
Thread Size 45
Color White
Flexibility Rigid