missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HTR1D Polyclonal Antibody
GREENER_CHOICE

Product Code. 15344721
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15344721 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15344721 Supplier Invitrogen™ Supplier No. PA533295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey, Panda, Dog (94%) Horse (88%) Marmoset, Bat, Rabbit, Guinea pig (81%).

G-protein coupled receptor for serotonin; found in nociceptors throughout the body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR1D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene HTR1D
Gene Accession No. P28221
Gene Alias 5 - Hydroxytryptamine (serotonin) receptor 1D; 5HT1D; 5-HT1D; 5-HT-1D; 5HT1D Receptor; 5-HT1d receptor; 5-HT-1D-alpha; 5-HT1DR; 5-hydroxytryptamine (serotonin) receptor 1D; 5-hydroxytryptamine (serotonin) receptor 1D beta; 5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled; 5-hydroxytryptamine receptor 1D; AI853647; Gpcr14; G-protein coupled receptor 14; HT1d receptor; HT1DA; Htr1d; HTR1DA; Htr1db; HTRL; RDC4; RP23-32L5.3; serotonin 1D alpha receptor; Serotonin receptor 1D
Gene Symbols HTR1D
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human 5HT1D Receptor.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3352
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.