missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR2B Polyclonal Antibody

HTR2B Polyclonal Antibody
Brand: Invitrogen PA533303
This item is not returnable.
View return policy
Description
HTR2B Polyclonal Antibody for IHC (P)
Specifications
| HTR2B | |
| Polyclonal | |
| Unconjugated | |
| HTR2B | |
| 5-HT 2B receptor; 5-HT(2B); 5-HT2B; 5-HT-2B; 5HT2B Receptor; 5-HT2b receptor; 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; AJ012488; AV377389; HTR2B; NP75 protein; serotonin receptor 2B; Srl; Stomach fundus serotonin receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P41595 | |
| HTR2B | |
| Synthetic 15 amino acid peptide from 2nd extracellular domain of human 5HT2B Receptor. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction